ContactUsLinkComponent
CRITERION™ Dehydrated Culture Media, Barney Miller Broth with Indicator, Hardy Diagnostics
CRITERION™ Dehydrated Culture Media, Barney Miller Broth with Indicator, Hardy Diagnostics
# de catalogue 75832-478
Fournisseur:  Hardy Diagnostics
Product available on GSA Advantage®
Numéro CAS:  
Barney Miller Broth with Indicator 2kg
CRITERION™ Dehydrated Culture Media, Barney Miller Broth with Indicator, Hardy Diagnostics
# de catalogue 75832-478
Fournisseur:  Hardy Diagnostics
Product available on GSA Advantage®
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Barney Miller Broth with Indicator
  • Size:
    2 kg
  • Cat. no.:
    75832-478
  • Supplier No.:
    C8992
Spécifications
A propos de cet article
Hardy Diagnostics CRITERION™ Barney Miller Broth with Indicator is recommended for the isolation and detection of beer spoilage microorganisms, specifically lactic acid bacteria
  • Wide mouth opening reduces dust formation by allowing the easy entry of a scoop
  • Convenient handgrip enables 'no slip' finger holds
  • Stackable containers for efficient storage
  • Increased solubility; dissolves quickly
  • Custom and bulk products available to meet customer specifications
  • Complete technical/instruction sheets, available in HUGO software
  • Unparalleled performance; tested for optimal growth
CRITERION™ Dehydrated Culture Media by Hardy Diagnostics is formulated to meet or exceed the highest standards. This broth is supplied in a 2kg bucket with convenient screw lid.
Indicator allows for early detection of growth by yellow color change due to pH shift.
Customers Who Bought This Also Bought