Spécifications
- Packaging:1 x 50 disks
- Description:XV Factor HardyDisks™, Hemin and NAD Disk
- Cat. no.:75832-734
- Supplier No.:Z7051
Spécifications
A propos de cet article
XV Factor HardyDisks™, Hemin and NAD disk is designed to aid in the identification of Haemophilus spp.
Contains NAD and Hemin (X and V Factors) for the speciation of Haemophilus. Each cartridge contains 50 disks that are spring loaded for smooth and reliable dispensing from the single metal disk dispenser by BD BioSciences. The Hardy Diagnostics manufacturing facility and quality management system comply with the FDA's Good Manufacturing Practices (GMP), and have achieved ISO 13485 certification as a medical device manufacturer.