Plasmodium falciparum, Gametocytes , Thick Smear Slide
# de catalogue 470181-954
Fournisseur: Avantor
Spécifications
- Description:Plasmodium falciparum, Gametocytes , Thick Smear
- Cat. no.:470181-954
- Stain:Giemsa
- Supplier No.:WARD470181-954
- Orientation:Smear
Spécifications
A propos de cet article
Can Cause Malaria in Humans
- Identifying features clearly distinguishable
- Rigorous quality control standards
Microscope slide showing the parasitic protozoan Plasmodium falciparum in human blood.
To learn more about this organism, please visit the CDC's article about the stages of Plasmodium falciparum in blood: https://www.cdc.gov/dpdx/resources/pdf/benchAids/malaria/Pfalciparum_benchaidV2.pdf
Customers Who Bought This Also Bought