RS-P25 Printer, METTLER TOLEDO®
# de catalogue CA97016-768
Fournisseur: Mettler Toledo
Spécifications
- Description:RS-P25 Printer
- Cat. no.:CA97016-768
- Supplier No.:11124320
- Paper Type:Standard paper
- Electrical:100–240V, 50/60Hz
- Interfaces:RS232C, 9 pin male connector
- Print speed:2.3 lines per sec.
Spécifications
A propos de cet article
The RS-P25 Compact Printer provides simple printouts on lightfast or adhesive paper.
- Fast and reliable printer
- User friendly operation
- RS232 interface
For use with certain METTLER TOLEDO® balances, titrators, and Ph meters.
Customers Who Bought This Also Bought