ContactUsLinkComponent
Disappearing Rainbow Demonstration
Disappearing Rainbow Demonstration
# de catalogue 470221-810
Fournisseur:  Ward's Science
Disappearing Rainbow Chemistry Demonstration kit
Disappearing Rainbow Demonstration
# de catalogue 470221-810
Fournisseur:  Ward's Science
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Disappearing Rainbow Demonstration
  • Cat. no.:
    470221-810
  • Supplier No.:
    IS7039
  • No. of Uses:
    7 demos
Spécifications
A propos de cet article
Demonstrate the concepts of Indicators and Acids and Bases!
  • Students see reactions in real time
  • Combine colors to create all the colors of the rainbow
Three indicators are used in conjunction with acids and bases to create a rainbow of colors. In a basic solution, phenolphthalein is red, p-nitrophenol is yellow, and thymolphthalein is blue. If done correctly the students only see the acid or base being poured, which adds to the impressive display. There are enough materials to complete the demonstration 7 times.
Ordering information: Materials include Red Indicator, Orange Indicator, Green Indicator, Blue Indicator, Violet Indicator, Hydrochloric Acid 1.0N, Sodium Hydroxide 1.0N, Sodium Hydroxide 6.0M, Instruction Manual. Material Needed but not included: (6) Glass Beakers or Clear Plastic Cups (at least 250mL), Graduated Cylinder, 100mL Distilled or Deionized water. Materials needed but not included are (6) Glass beakers or clear plastic cups (at least 250ml), graduated cylinder, 100ml, distilled or deionized water.
Delivery: Due to the nature of this item, it cannot be shipped by air under any circumstances. Please select ground shipping at time of checkout.
Caution: Students see reactions in real time Combine colors to create all the colors of the rainbow
Customers Who Bought This Also Bought