FireKing® Turtle® Vertical File Cabinets, Essendant LLC MS
Fournisseur: Fireking International Inc
Certificats
A propos de cet article
This is made of 100% gypsum, reinforced by a 1" by 2" lattice made of 14-gauge galvanized welded steel wire, providing complete peace of mind from fire, impact, or explosion of over 1700 ˚F proves the Turtle is built to survive under the worst conditions.
- Letter- and legal-sized filing
- Recessed handles
- Replaceable panels
Oven-dried, asbestos-free insulation maintains the protection for a lifetime. Environmental-friendly electrostatic powder coating applied to all sides, including bottom.
Because of its unique steel lattice reinforcement, insulation between drawers and extra-thick steel panels, the Turtle survive a loaded fall of 30' and still
keep its contents safe in a fire.
Customers Who Bought This Also Bought