Western Blotting Kit, Azure Biosystems
Fournisseur: Azure Biosystems
Certificats
A propos de cet article
Azure's fluorescent western kits provide reagents and materials for two-color fluorescent Western blots.
- Pre-cut membranes
- Wash buffer
- Visualize two proteins by fluorescence
Western Blotting Kit for visualizing two proteins by fluorescence. Enough for 10 Western blots. Each kit includes pre-cut membranes, wash buffer, fluorescent blot blocking buffer, quenching sheets, and secondary antibodies.
Customers Who Bought This Also Bought