Maintenance-Free Tyvek® Hood Assembly with Low Pressure Flow Adapter and Suspension, Allegro
# de catalogue 89011-732
Fournisseur: Allegro
Small Business Enterprise
Spécifications
- Garment application:Laboratory
- Cat. no.:89011-732
- Supplier No.:9910
Spécifications
A propos de cet article
Our NIOSH-approved Fully Disposable Tyvek Hood is so unique, it is patented.
- Poly-Coated DuPont Tyvek Hood
- Splash-Resistant Hood and Bib
- Seams Surged to Seal in The Air Flow
- Generous Size is Ideal for Beards or Goggles
- Low-Profile, Lightweight Flow Adapter
It’s designed and constructed in one piece with the down tube and head suspension completely integrated
The Low Pressure Flow Adapter allows the hood to connect to an ambient air pump or any low pressure air source. It's economical enough to dispose of with the hood assembly or save and use with a replacement hood.
This makes it maintenance free and eliminates the need for decontamination of the breathing tube or replacement parts. The entire assembly is disposed of after use.
Pressure range: 6 to 43 psi
Customers Who Bought This Also Bought