Disposable Large Respirator Storage Bags, Allegro
# de catalogue 47747-250
Fournisseur: Allegro
Small Business Enterprise
Spécifications
- Ø:8"
- W×L:11×21"
- Cat. no.:47747-250
- Supplier No.:4001-05
Spécifications
A propos de cet article
Disposable storage bags with gussets help prevent cross contamination of any personal safety equipment such as respirators or ear muffs.
- Pressure-sensitive seals for recording cleaning information
- Single-Use
The large bags come with pressure-sensitive seals where cleaning information can be recorded.
Customers Who Bought This Also Bought