ECO Containers for ECO Funnels, CP Lab Safety
Fournisseur: CP Lab Safety
Certificats
A propos de cet article
ECO Funnels® are designed to fit a wide variety of standard Nalgene® carboys, Justrite® poly waste cans and other containers. The selection below includes the most common containers used with the ECO Funnel laboratory.
- Made from PE
- Good chemical resistance when glass is not an option
Containers with 38 mm, 53 mm, and 70 mm openings will typically work with one of the ECO Funnels.
Customers Who Bought This Also Bought