ContactUsLinkComponent
Regan-Lowe Agar, Hardy Diagnostics
Regan-Lowe Agar, Hardy Diagnostics
# de catalogue 10142-316
Fournisseur:  HARDY DIAGNOSTICS CA
Product available on GSA Advantage®
Numéro CAS:  
REGAN-LOWE AGAR 15X100 PLATE PK 10
Regan-Lowe Agar, Hardy Diagnostics
# de catalogue 10142-316
Fournisseur:  HARDY DIAGNOSTICS CA
Product available on GSA Advantage®
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Regan-Lowe Agar, 15x100mm Plate
  • Size:
    22 ml
  • Cat. no.:
    10142-316
  • Supplier No.:
    A65
Spécifications
A propos de cet article
Charcoal Blood Agar with 10% Horse Blood and Cephalexin is a selective medium recommended for the isolation and cultivation of Bordetella pertussis.
The basal medium of Regan-Lowe Agar consists of charcoal agar supplemented with defibrinated horse blood. Charcoal, along with starch, neutralizes fatty acids and peroxides, which are toxic to Bordetella. Horse blood is an added enrichment which supports the growth of Bordetella spp. Cephalexin inhibits the growth of normal flora of the nasopharynx. Yeasts and fungi are inhibited by the inclusion of amphotericin B. Beef extract and enzymatic digest are incorporated in the medium to supply amino acids and other nitrogenous substances that are necessary for bacterial growth. Osmotic equilibrium is maintained by the addition of sodium chloride. Niacin (nicotinic acid) is a vitamin which is added for growth promotion.
Certifications: ATCC No.9797
Customers Who Bought This Also Bought