A propos de cet article
C-Flex® is a unique, patented thermoplastic elastomer specifically designed to meet the critical demands of the pharmaceutical, research, biopharmaceutical and diagnostics industries. C-Flex® biopharmaceutical tubing has been used by many of the world’s leading biotech and pharmaceutical processing companies.
- Non-pyrogenic, non-cytotoxic, non-hemolytic
- Chemically resistant to concentrated acids and alkalis
- Low platelet adhesion, low protein binding
- Ultra-smooth inner bore
- Available in clear or opaque formulations
- Lot traceable, safe disposal through incineration
Each coil of C-Flex tubing is extruded to precise Int.Ø, Ext.Ø.
Temperature resistance: –45 to +135 °C
Hardness (Shore durometer A): 59
Tensile strength, psi: 900
Elongation, %: 740
Tensile modulus, @100%/300%, psi: 240/400
Tensile set @ 300% Stretch: 19
Graves tear strength, pli Die C: 150
Compression set constant defl., 'B' (22 hrs. @ 70 °C), %: 53
Water absorption, % at 100% RH: 0.017
Brittle point: –60 °C
Low temperature flexibility @ –40 °C: Pass
All tubing is formulated to meet the standards Eachof the biopharmaceutical industry and is QA tested before leaving the Saint-Gobain performance plastics production facility.