ContactUsLinkComponent
Difco™ TCBS (Thiosulfate-Citrate-Bile-Sucrose) Agar, BD Biosciences
Difco™ TCBS (Thiosulfate-Citrate-Bile-Sucrose) Agar, BD Biosciences
# de catalogue CA90003-770
Fournisseur:  BD
Numéro CAS:  
Difco™ TCBS (Thiosulfate-Citrate-Bile-Sucrose) Agar, BD Biosciences
# de catalogue CA90003-770
Fournisseur:  BD
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Difco™ TCBS (Thiosulfate-Citrate-Bile-Sucrose) Agar
  • Size:
    500 g
  • Cat. no.:
    CA90003-770
  • Supplier No.:
    265020
Spécifications
A propos de cet article
TCBS Agar is used for the selective isolation of cholera vibrios and Vibrio parahaemolyticus from a variety of clinical and nonclinical specimens.
  • Oxgall inhibits the growth of Gram-positive bacteria
  • Sodium thiosulfate serves as a sulfur source
  • Sodium thiosulfate detects hydrogen sulfide production
  • Sucrose is included as a source of carbohydrates
  • Alkaline pH enhances the recovery of V. cholerae
Thymol blue and bromthymol blue are included as indicators of pH changes.
Customers Who Bought This Also Bought