Order Entry
ContactUsLinkComponent
BBL™ SFP Agar Base, BD
BBL™ SFP Agar Base, BD
# de catalogue CA90004-026
Fournisseur:  BD
Numéro CAS:  
BBL™ SFP Agar Base, BD
# de catalogue CA90004-026
Fournisseur:  BD
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Spécifications

  • Description:
    BBL™ SFP Agar Base
  • Size:
    500 g
  • Cat. no.:
    CA90004-026
  • Supplier No.:
    281110

Spécifications

A propos de cet article

SFP Agar Base is used with Egg Yolk Enrichment 50%, Antimicrobic Vial P and Antimicrobic Vial K in detecting and enumerating Clostridium perfringens in foods.

  • 50% egg yolk enrichment
  • Dehydrated appearance: Beige, free-flowing
  • Peptones provide carbon, nitrogen, vitamins, and minerals
  • Agar is the solidifying agent

The selectivity of the medium is due to the added kanamycin and polymyxin B.

Customers Who Bought This Also Bought