SP Wilmad-LabGlass Silicone Rubber Septum with PTFE Coating on One Side, SP Industries
# de catalogue 14008-020
Fournisseur: SP Industries
Spécifications
- Ø:20-400
- Septum:PTFE/Silicone
- Description:Septum
- Thickness:0.065" (0.005 PTFE and 0.065 silicone)
- Cat. no.:14008-020
- Supplier No.:LG-4342-108
Spécifications
A propos de cet article
PTFE face provides an inert contact surface while the silicone backing material effects a positive seal
Frequently used when syringe techniques are indicated.
Customers Who Bought This Also Bought