Standard-Incubators with Natural Convection, Avantgarde.line BD Series, BINDER
Fournisseur: Binder
Certificats
Ć propos de cet article
The BD Avantgarde.Line series is specially designed for gentle incubation of organisms, as well as conditioning of heatsensitive media. The design of the units and the homogeneous temperature distribution ensure the very best results.
- Excellent temporal and spatial temperature accuracy with high process reliability
- Disinfection routine of 100 °C
- High temperature accuracy with APT.line⢠technology
- Controller with LCD display
- Electromechanical control of the exhaust air flap
- Ergonomic handle design
- USB port for recording data
Features inner door made of tempered safety glass with two chrome-plated racks. Class 3.1 integrated independent temperature safety device (DIN 12880) is available with visual alarm.
Units up to 115 L are stackable.
SpƩcifications
- Catalog No:
- 75783-558
- 75783-618
- 75783-554
- Convection type:
- Natural convection
- Natural convection
- Natural convection
- Temperature range:
- Ambient +5ā¦100 °C
- Ambient +5ā¦100 °C
- Ambient +5ā¦100 °C
SpƩcifications
Informations connexes
Services: VWRCATALYST⢠offers validation services including IQ, OQ, and PQ protocol development and execution, as well as asset management and calibration services (US only); call 888-793-2300 for more details.