Drying/Heating Ovens with Natural Convection, Avantgarde.line ED Series, BINDER
Fournisseur: BINDER INC MX
Certificats
A propos de cet article
BINDER Avantgarde.Line ED Drying ovens are ideal for sterilization, drying, and thermal storage tasks which do not require high drying rates or special time parameters.
- Ovens provide reliable, reproducible, quick task results
- Easy to clean
BINDER ovens provide fast uniform drying for drying and sterilization applications. The Advanced Preheating Technology (APT.line™) provides temperature accuracy of 1°C with a wide temperature range from 5°C to 15°C above ambient to 300 °C (depending on the model size). With the natural convection and high rate of air exchange, thermal process run with increased efficiency.
Certifications: UL.
Ordering information: The ovens include two chrome-plated shelves. For additional sizes not listed below, please refer to the BINDER Classic ED Drying Ovens.
Customers Who Bought This Also Bought