UltraShield UHPLC PreColumn Filter, Restek
Fournisseur: Restek
Certificats
A propos de cet article
Cost effective pre-column filters to protect UHPLC systems.
- Reliable way to extend column lifetime
- Universal fit
- Connects easily to any brand column
- Leak-tight to 15000 psi (1034 bar)
- 0.5 µm or 0.2 µm titanium frit in a stainless steel body
- PEEK ferrule
Customers Who Bought This Also Bought