Rhino VET 360 Veterinary Refractometer, Reichert
# de catalogue 51100-100
Fournisseur: Reichert Technologies
Spécifications
- Description:Rhino VET 360 Veterinary Refractometer
- Height:38 mm
- Model:VET 360
- Scales:Urine Specific Gravity: Small Animals,
Urine Specific Gravity: Large Animals,
Serum Protein - Cat. no.:51100-100
- Accuracy:Urine specific gravity: ±0.001, serum protein: ±0.2 g/100 ml
- Length:235 mm
- Range:Urine specific gravity: 1.000 - 1.060, serum protein: 2.0 - 14.0 g/100ml
- Weight:0.16 kg
- Width:38 mm
- Supplier No.:137536L0
Spécifications
A propos de cet article
The Rhino-rugged water resistant, dustproof, and shock resistant hand-held refractometer uses Automatic Temperature Compensation to provide accurate readings regardless of ambient temperature.
- Automatically corrects all readings back to the Standard Reference Temperature of 68°F (20°C) providing greater accuracy
- Improved optics that are easy to read
- No batteries required making it reliable any time
- Ideal for use in the office or outdoors
- Large and small animal scales in one instrument
- IP67 rated, sealed internal optics cannot be contaminated
- Test takes only seconds
Customers Who Bought This Also Bought