Sealing Films, Aeraseal 96, Chemglass
Fournisseur: Chemglass
Certificats
A propos de cet article
A 114 µm thick hydrophobic porous sealing film with medical-grade adhesive for tissue culture plates, bio-blocks, and 96-well plates where gas exchange is necessary for cell or bacterial growth
- Non-Cytotoxic, Highly Gas Permeable
- Easily Pierceable with Pipet Tips for Sample Recovery
- Recommended for Temperatures from -20° to +80°C
AeraSeal™ sealing films minimize cross-contamination, spillage and evaporation. They allow uniform air and CO2 exchange for all wells, unlike plate lids which favor exchange for the wells near the plate edges. Dimensions: 82.6 x 142.9 mm for standard-size tissue culture plates.
Customers Who Bought This Also Bought