ContactUsLinkComponent
Wave Apparatus
Wave Apparatus
# de catalogue 470061-736
Fournisseur:  VWR International
Wave Apparatus Demo - Metal
Wave Apparatus
# de catalogue 470061-736
Fournisseur:  VWR International
Spécifications
  • Description:
    Wave Apparatus
  • Cat. no.:
    470061-736
  • Supplier No.:
    PH0712
Spécifications
A propos de cet article
A Classic Demonstration!
  • Demonstrate longitudinal and transverse motion with ease
This advanced apparatus demonstrates both longitudinal and transverse wave motions. Consisting of a number of eccentric discs supporting a series of metal rods revolving on a handle, transverse waves are obtained. Longitudinal waves are obtained from the bent rods. The rotating handle is equipped with a circular scale marked in degrees to indicate rotation. Includes instructions.
Customers Who Bought This Also Bought