TE (Tris EDTA), 1X, pH 8.0 (Nuclease Free)
Fournisseur: GROWCELLS
Certificats
A propos de cet article
Nuclease free TE is used to solubilize DNA/RNA while providing protection from degradation
It provides great buffering capabilites for electrophoresis, and can be used for general storage of nucleic acids.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.