3M Sodium Acetate Solution, pH 5.5
Fournisseur: GROWCELLS
Certificats
A propos de cet article
3M Sodium acetate.
Ready-to-use sterile solution made with 18 megOhm water, 0.2 µm sterile filtered, pH adjusted to 5.5 with glacial acetic acid. Ideal for DNA purification.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.