ContactUsLinkComponent
Egg Yolk Agar, Modified, Hardy Diagnostics
Egg Yolk Agar, Modified, Hardy Diagnostics
# de catalogue 75813-978
Fournisseur:  Hardy Diagnostics
Product available on GSA Advantage®
Numéro CAS:  
Egg Yolk Agar, Modified, for Clostridium, 18ml fill, 15x100mm plate
Egg Yolk Agar, Modified, Hardy Diagnostics
# de catalogue 75813-978
Fournisseur:  Hardy Diagnostics
Product available on GSA Advantage®
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Format:
    15×100 mm plate
  • Description:
    Egg Yolk Agar, Mod.
  • Size:
    Pack of 10
  • Minimum Order Quantity:
    10
  • Supplier No.:
    G215
Spécifications
A propos de cet article
Egg Yolk Agar, Modified, for Clostridium is used for the isolation, cultivation, and differentiation of Clostridium spp.
  • Isolation
  • Cultivation
  • Differentiation
For the isolation, cultivation, and differentiation of Clostridium spp. and other anaerobic bacteria.
Ordering information: Minimum order of 10 packs.
Customers Who Bought This Also Bought