Mueller Hinton Agar with Blood, Hardy Diagnostics
# de catalogue 75814-684
Fournisseur: Hardy Diagnostics
Product available on GSA Advantage®
Numéro CAS:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Format:15×150 mm plate
- Description:Mueller Hinton Agar w/Bld, HDx
- Supplier No.:H21
Spécifications
A propos de cet article
Mueller Hinton Agar with Blood is used for disk diffusion sensitivity testing of Streptococcus spp.
- Disk diffusion sensitivity training
- With 5% sheep blood
Mueller Hinton Agar with 5% Sheep Blood For disk diffusion sensitivity testing of Streptococcus spp.
Customers Who Bought This Also Bought