Blank Paper Disk for AST, Hardy Diagnostics
# de catalogue 75813-972
Fournisseur: Hardy Diagnostics
Product available on GSA Advantage®
Numéro CAS:
Spécifications
- Description:Blank Disk
- Size:0.25"
- Cat. no.:75813-972
- Supplier No.:Z7121
Spécifications
A propos de cet article
A blank paper disk that can be used for inhibition studies.
- Prepare your own disks for Kirby Bauer (antimicrobial disk diffusion (AST)) testing
- Used for inhibition studies
A blank paper disk that can be used for inhibition studies. Used to prepare your own disks for Kirby Bauer (antimicrobial disk diffusion (AST)) testing on Mueller Hinton Agar.
Customers Who Bought This Also Bought