Potato Flake Selective Agar (with Ciprofloxacin and Vancomycin) Deep Fill Plate, Hardy Diagnostics
# de catalogue 75814-672
Fournisseur: Hardy Diagnostics
Product available on GSA Advantage®
Numéro CAS:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Packaging:Deep Fill Plate
- Description:Potato Flake Selective Agar (with Ciprofloxacin and Vancomycin)
- Size:15 x 100 mm
- Cat. no.:75814-672
- Fill Volume:—
- Supplier No.:W159
Spécifications
A propos de cet article
Potato Flake Selective Agar is used for the selective isolation of pathogenic fungi.
Customers Who Bought This Also Bought