ContactUsLinkComponent
EnviroTrans™ Swab Rinse Kit, Barney Miller Broth, Hardy Diagnostics
EnviroTrans™ Swab Rinse Kit, Barney Miller Broth, Hardy Diagnostics
# de catalogue 75813-998
Fournisseur:  Hardy Diagnostics
Product available on GSA Advantage®
Numéro CAS:  
EnviroTrans™, swab rinse kit, Barney Miller Broth, 5mL without swab
EnviroTrans™ Swab Rinse Kit, Barney Miller Broth, Hardy Diagnostics
# de catalogue 75813-998
Fournisseur:  Hardy Diagnostics
Product available on GSA Advantage®
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Barney Miller Broth
  • Cat. no.:
    75813-998
  • Supplier No.:
    SRK100
Spécifications
A propos de cet article
EnviroTrans™ Swab Rinse Kit, Barney Miller Broth is recommended for environmental monitoring.
  • Collect and transport environmental samples with efficiency and ease.
  • Prefilled with Barney Miller Broth
Recommended for environmental monitoring.
Collect and transport environmental samples with efficiency and ease. Simply remove the cap and attached Dacron® swab, collect the sample, recap, and send to the lab. Each EnviroTrans™ tube contains broth or buffer.
Customers Who Bought This Also Bought