VWR® 25D Digital Homogenizer
Catalog # 75800-448
Supplier: VWR International
Specifications
- Description:VWR®25D Digital Homogenizer
- Sound level:68 dB
- Dimensions:35.56 × 30.48 × 60.96 cm (14 × 12 × 24")
- Power rating:576 W
- Weight:8.16 kg (18 lbs.)
- Cat. no.:75800-448
- Electrical:120 V
- Speed Range:0 - 30000 rpm
- Volume Range:0.05 ml - 3.5 L
Specifications
About this item
The versatile and powerful VWR® 25D Homogenizer is ideal for a wide range of lab applications, including but not limited to compounding, emulsions and suspensions, cell disruption, protein extractions, medical research or pharmaceutical processing.
- Compact digital homogenizer unit with high watt motor
- 0.05ml - 3.5 L volume range and viscosity up to 50,000 mPas
- Bright LED display and user-friendly controls
- Can homogenize, emulsify, blend and/or mix
- Includes U-shape post mounted stand assembly
- VWR® Generator probe sold separately
The VWR® 25D Homogenizer has a wide volume capability with processing up to 3.5 Liters (higher sample volumes dependent on viscosity). Its digital display and speed control provides the ability to fine-tune speeds between 0 to 30,000 RPM, allowing its speed adjustment to be precisely controlled within the range.
Certifications: CE marked.
Accessories information: Generator probes must be purchased with homogenizer units.
Ordering information: Supplied with a two-year manufacturer's warranty. VWR® Generator probe(s) are required for use with the VWR25D Homogenizer model and are sold separately (VWR® GenPack Adapter is required for VWR® GenPack Generator Probes).