VWR® Spatulas, PTFE-Coated
Supplier: VWR International
Certificates
About this item
VWR® PTFE-Coated Spatulas are available in a variety of styles to accommodate an array of laboratory tasks.
- Passivated
- Compatible with Gamma/ETO/Autoclavable sterilization methods
All spatulas feature stainless steel construction, with a PTFE coating.
Customers Who Bought This Also Bought