VWR® Cartilage Knife
# de catalogue 10806-332
Fournisseur: VWR International
Spécifications
- Length:216 mm (8.5")
- Description:VWR® Cartilage Knife
- Material:Stainless steel/wood
- Blade Length:89 mm
Spécifications
A propos de cet article
The VWR® Cartilage Knife features a high-quality, stainless steel blade securely attached to a riveted wooden handle
- Passivated
- Compatible with Gamma/ETO/Autoclavable sterilization methods
Blade Length is 89mm (3.5").
Customers Who Bought This Also Bought