VWR® Petri Dish Sterilization Box
Catalog # 10806-404
Supplier: VWR International
Specifications
- Description:VWR® Petri Dish Sterilization Box
- Dimensions:108×267 mm (4.25×10.5")
- Cat. no.:10806-404
Specifications
About this item
The VWR® Petri Dish Sterilization Box is made from polished ASTM standard (899-94) rugged stainless steel.
- Passivated
- Compatible with Gamma/ETO/Autoclavable sterilization methods
Each box accommodates 18 glass Petri dishes sized 100x10mm (3.93×0.39"), or 10 dishes sized 100x15mm (3.93×0.59").
Customers Who Bought This Also Bought