VWR® Tin Specimen Box
Fournisseur: VWR International
Certificats
A propos de cet article
VWR® Tin Specimen Boxes are manufactured via an ASTM standard (A623-11) Tin-Milling process
- Passivated
- Round, seamless tin construction
- Low-form, flat bottom design
- Supplied with label on lid
- Compatible with Gamma/ETO/Autoclavable sterilization methods
Boxes are excellent for sample collection and storage.
Customers Who Bought This Also Bought