VWR® Pure Nickel Dish
Catalog # 10806-390
Supplier: VWR International
Specifications
- Capacity:0.134 L (4.5 oz.)
- Description:VWR® Pure Nickel Dish
- Dimensions:70 O.D. x 35 H mm (2.75 x 1.375")
- Cat. no.:10806-390
Specifications
About this item
The VWR® Pure Nickel Dish is made from a one-piece ASTM standard (A-494) Pure Nickel sheet, spun to shape
- Passivated
- Compatible with Gamma/ETO/Autoclavable sterilization methods
Dishes feature straight sides, with a lip for pouring.
Customers Who Bought This Also Bought