DataChart® 1250 Two Channel Paperless Recorder, Monarch
Fournisseur: Monarch Instrument
Certificats
A propos de cet article
The DataChart® 1250 is a feature-rich digital recording system offering 2 universally configurable inputs for measuring DC voltage, DC current, thermocouples, and RTDs, as well as frequency and pulse inputs
A maximum sample storage rate of 100 samples per second can be set for both channels, allowing for the capture of short duration process signal anomalies. CompactFlash™ cards up to 2 Gigabyte size can be used, allowing many data points to be stored over long periods of time.
Standard features include 4 internal alarm setpoints, 2 alarm relay outputs, and 1 digital control input. Combining small package size with extreme flexibility, the DataChart® 1250 is a powerful, yet economical digital data recording system.
Certifications: CE compliant. RoHS compliant.
Ordering information: Units include a manufacturer's one-year warranty, and Navigtor Lite Software for exporting data.
Customers Who Bought This Also Bought