Heratherm® Drying Ovens, Advanced Protocol Security Series, Thermo Scientific
# de catalogue 10124-590
Fournisseur: Thermo Fisher Scientific
Spécifications
- Temperature Range:50°C - 330°C
- Volume:62 L (2.2 cu. ft.)
- Description:Security Oven, Stainless Steel Exterior
- Interior Dimensions:35.4 x 34.3 x 50.8 cm (13.9 x 13.5 x 20")
- Exterior Dimensions:53 x 56.5 x 72 cm (20.9 x 22.2 x 28.3")
- Electrical:120 V, 60 Hz, 1440 W, 12 A
- No. of Shelves:13 max.
- Weight:97 lbs.
- Maximum Load per Shelf:55 lbs.
- Supplier No.:51028539
Spécifications
A propos de cet article
Heating and drying oven with stainless steel exterior.
- Robust and corrosion-resistant surface
- Standard over-temperature alarm and additional under-temperature alarm
- Programmable controller for temperature ramps and dwells saves up to 10 programs
- Access port allows the introduction of sensors for independent data monitoring
- Boost function enables rapid heat up
- Auto-dry function deactivates oven when samples are dry (with use of optional temperature sensor)
- Lockable door with door alarm
The timer allows the oven to be turned on or off after a preset time, with a choice between weekly, hourly or real time settings. Units with natural convection are ideal for gentle drying and heating, to protect delicate samples. Units with forced convection have adjustable fan speed and are suitable for faster drying and heating, and superior temperature uniformity.
Delivery: Supplied with two shelves and standard RS232 data interface.
Customers Who Bought This Also Bought