VWR® Mixter Forceps
Fournisseur: VWR International
Certificats
A propos de cet article
VWR® Mixter Forceps are constructed of surgical-grade stainless steel.
- Passivated
- Box-lock joint
- Fully autoclavable
- Full serrated jaws
- Rockwell hardness (HRC): 40–45
These forceps can be sterilized by EtO/gamma radiation.
Customers Who Bought This Also Bought