VWR® Rankin Kelly Hemostat Dissecting Forceps
Supplier: VWR International
Certificates
About this item
VWR® Rankin Kelly Hemostat Dissecting Forceps are constructed of surgical-grade stainless steel.
- Passivated
- Box-lock joint
- Fully autoclavable
- Half-serrated jaws
- Rockwell hardness (HRC): 40–45
These forceps can be sterilized by EtO/gamma radiation.
Customers Who Bought This Also Bought