VWR® Premium Kelly Hemostat Dissecting Forceps
Supplier: VWR International
Certificates
About this item
VWR® Premium Kelly Hemostat Dissecting Forceps are constructed of German steel.
- Passivated
- Box-lock joint
- Fully autoclavable
- Half-serrated jaws
- Rockwell hardness (HRC): 40–45
These reusable forceps can be sterilized by EtO/Gamma radiation.
Customers Who Bought This Also Bought