3510 White LD-100 Frock, Worklon®
Fournisseur: Superior Uniform
Certificats
A propos de cet article
LD-100 polyester taffeta frocks are often used in pharmaceutical, medical device, food processing, and related industries where particulate control is required, and garments are donned in controlled environments up to and including Class 1000 (ISO 6)
The frocks feature a snap closure, raglan sleeves, polyester knit cuffs, two-ply reinforced placket, adjustable snaps at collar, and a color-coded hang loop denoting size.
Spécifications
- Catalog No:
- 89136-770
- 89136-772
- 89136-778
- 89136-780
- 89136-774
- 89136-776
- 89136-782
- 89136-784
- Model:
- 3510
- 3510
- 3510
- 3510
- 3510
- 3510
- 3510
- 3510
- Garment application:
- Medical, Food, Laboratory
- Medical, Food, Laboratory
- Medical, Food, Laboratory
- Medical, Food, Laboratory
- Medical, Food, Laboratory
- Medical, Food, Laboratory
- Medical, Food, Laboratory
- Medical, Food, Laboratory
- Gender:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Closure type:
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Fabric:
- Polyester
- Polyester
- Polyester
- Polyester
- Polyester
- Polyester
- Polyester
- Polyester