ContactUsLinkComponent
Round Cover Glass, Azer Scientific
Round Cover Glass, Azer Scientific
# de catalogue 89187-782
Fournisseur:  Azer Scientific Inc.
Product available on GSA Advantage®
Minority or Woman-Owned Business Enterprise
COVER GLASS 12MM RND #1 PK100
Round Cover Glass, Azer Scientific
# de catalogue 89187-782
Fournisseur:  Azer Scientific Inc.
Product available on GSA Advantage®
Minority or Woman-Owned Business Enterprise
Spécifications
  • Ø:
    12 mm
  • Numbering:
    #1
  • Description:
    Cover glasses, round
  • Supplier No.:
    200121
Spécifications
A propos de cet article
The round cover glass is pure white borosilicate glass of the first hydrolytic class.
  • Excellent chemical resistance
  • Optimal flatness
  • No disrupting fluorescence
Cover glass does not emit any disturbing fluorescence under the ultraviolet light of the fluorescence microscope.
Customers Who Bought This Also Bought