Life Latch® New Generation Pails and Lids, M&M Industries
Fournisseur: M&M Industries
Certificats
A propos de cet article
User friendly screw-top pail for ultimate product protection.
- Moisture-resistant, air-tight seal
- White screw-top pail supplied with white gasketed lid
- FDA grade raw materials
- 8 color offset printing
- Tamper evident
- Patented "no gasket cover" allows products to breathe
Wall thickness: 0.05 - 0.09"
Certifications: UN certified for solid groups II and III.
Customers Who Bought This Also Bought