Spécifications
- Length:38 mm (11/2")
- Tip Configuration:Solid Blades
- Curvature:Straight
- Cat. no.:10841-340
- Supplier No.:65-1760
Spécifications
A propos de cet article
This is an ophthalmic instrument used to retain the eyelids open, often during cataract or phacoemulsification procedures.
- Made of stainless steel
It is utilized to provide the surgeon with increased visualization and access to the surgical site. This speculum is lightweight and does not put a great deal of pressure on the globe. To a degree, the surgeon can increase the tension of the instrument by manipulating the wire where the speculum is held, as it is slightly maleable. In situations where a wire speculum will not provide enough force to open the lids sufficiently, a speculum with a screw mechanism may be used.