Spécifications
- Length:44 mm (13/4")
- Sterility:Non sterile
- Disposable / reusable:Reusable
- Latex/Latex-free:Latex-free
- Material:Stainless Steel
- Tip Configuration:Blunt
- Curvature:Angled
- Supplier No.:65-1754
Spécifications
A propos de cet article
The Kratz-Barraquer Wire Speculum is an ophthalmic instrument used to retain the eyelids open, often during cataract or phacoemulsification procedures
- Light weight
It is utilized to provide the surgeon with increased visualization and access to the surgical site. To a degree, the surgeon can increase the tension of the instrument by manipulating the wire where the speculum is held, as it is slightly maleable. In situations where a wire speculum will not provide enough force to open the lids sufficiently, a speculum with a screw mechanism may be used.