Spécifications
- Material:Stainless Steel
- Curvature:Slightly Curved
- Cat. no.:10197-860
- Length:95 mm (3 3/4")
- Supplier No.:64-1348
- Blade Type:Sharp/Sharp
Spécifications
A propos de cet article
The Reeh Stitch Scissors are used for suture removal
These scissors have short blades with sharp, fine tips that have a very small hook-shaped tip on one blade that slides under sutures to slightly lift them before cutting for removal. These Stitch Scissors should not be used for cutting tissue. They also feature Ribbon Style Finger Ring handles for increased comfort and improved grip. Reeh Stitch Scissors are often used for removing sutures from Ophthalmic, Dental, Plastic, and Reconstructive surgeries.