ContactUsLinkComponent
Reeh Stitch Scissors, OR Grade, Sklar
Reeh Stitch Scissors, OR Grade, Sklar
# de catalogue 10197-860
Fournisseur:  Sklar
Scissors
Reeh Stitch Scissors, OR Grade, Sklar
# de catalogue 10197-860
Fournisseur:  Sklar
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Material:
    Stainless Steel
  • Curvature:
    Slightly Curved
  • Cat. no.:
    10197-860
  • Length:
    95 mm (3 3/4")
  • Supplier No.:
    64-1348
  • Blade Type:
    Sharp/Sharp
Spécifications
A propos de cet article
The Reeh Stitch Scissors are used for suture removal
These scissors have short blades with sharp, fine tips that have a very small hook-shaped tip on one blade that slides under sutures to slightly lift them before cutting for removal. These Stitch Scissors should not be used for cutting tissue. They also feature Ribbon Style Finger Ring handles for increased comfort and improved grip. Reeh Stitch Scissors are often used for removing sutures from Ophthalmic, Dental, Plastic, and Reconstructive surgeries.
Customers Who Bought This Also Bought