Ovation® Fixed Volume Pipettors, VistaLab
Supplier: VistaLab
Certificates
About this item
Mechanical 1-stroke (F1) or 2-stroke (F2) pipettors with low-force plunger operation.
- Ovation F1 models available in 11 fixed volumes ranging from 5 to 1000 μl
- Ovation F2 models available in 13 fixed volumes ranging from 1 to 1000 μl
- Built-in microtube cap opener
- Custom fixed volumes available upon request
Ovation pipettors and essential components are available, with pipettors featuring mechanical low-pressure plunger operation.
Customers Who Bought This Also Bought