MLA Precision Pipettors, VistaLab
Fournisseur: VistaLab
Certificats
A propos de cet article
MLA pipettors are made of precision-engineered aluminum alloy and stainless steel, and are designed for highly accurate delivery and proven performance.
- Available in a wide range of fixed volumes
- Light plunger forces for easy operation with thumb/forefinger
- Single-stroke 'to deliver' operation
- Individually serialized
- Lifetime warranty on parts
These pipettors are available in 28 fixed volumes ranging from 5 to 1000 μL. Pipettors can be easily adjusted as needed to achieve accurate performance on dense or viscous liquids.
BodyTextAccessories: Seal Kits for MLA Pipettors are available separately. Seal kits contain lubricant, seals, and instructions.
Customers Who Bought This Also Bought