Thermocouple Adapter, Chemglass
Fournisseur: Chemglass
Certificats
A propos de cet article
These thermocouple adapters provide quick and easy connections between standard (OST) and miniature (SMP) connector systems.
- Quick and easy
- Provides connection between OST and SMP systems
Customers Who Bought This Also Bought