Life Latch® Screw Top UN Rated Poly Pail, New Pig
# de catalogue 10218-578
Fournisseur: New Pig
Spécifications
- Capacity:4.7 L (1.25 gal.)
- Description:Life Latch® Screw top UN rated poly pail, 8.87x9.14"
- Cat. no.:10218-578
- Supplier No.:DRM1048
Spécifications
A propos de cet article
Store and transport solids with confidence in these UN Rated pails.
- Built-in locking mechanism
- Open-head container
- Can also be used for storing liquids
Store and transport solids with confidence in these UN Rated pails. Just pinch the latches to unlock and remove lid! Built-in locking mechanism lets you close lid simply and securely. Open-head container is UN Rated for storing or transporting solid materials; can also be used for storing liquids. Durable HDPE construction is chemical resistant and FDA Approved. Slot on lid accepts cable ties (not included) for applications that require a tamper-proof pail. Attached handle makes carrying and loading easier.
Customers Who Bought This Also Bought