ContactUsLinkComponent
Muscle (Mammalian), 3 Types Slide
Muscle (Mammalian), 3 Types Slide
Catalog # 470177-586
Supplier:  Avantor
MUSCLE (MAMMALIAN) 3 TYPES SECT
Muscle (Mammalian), 3 Types Slide
Catalog # 470177-586
Supplier:  Avantor
Specifications
  • Description:
    Muscle (Mammalian), 3 Types Slide
  • Cat. no.:
    470177-586
  • Stain:
    Hematoxylin and Eosin
  • Supplier No.:
    WARD470177-586
  • Orientation:
    Section
Specifications
About this item
Compare and contrast muscle types
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Mammal. Entire small muscle.
Customers Who Bought This Also Bought